EMAIL THIS PAGE TO A FRIEND
Genes & development
Creating cell type-specific mutants by enhancer mutagenesis.
PMID 28446593
Abstract
Cell signaling plays an essential role in development, and knowledge of the identities of the cells sending the signal is critical. This can be a challenge, since signaling pathways and ligands are commonly used at multiple times and in multiple cell types during development. One solution to this problem is to create cell type-specific mutants using CRISPR/Cas9 to mutate enhancers that control different patterns of expression. In this issue of Genes & Development, Rogers and colleagues (pp. 634-638) provide the first use of this method in Drosophila to solve a long-standing issue in patterning of the embryonic central nervous system.