EMAIL THIS PAGE TO A FRIEND
Food chemistry
Encapsulation of fish oil into hollow solid lipid micro- and nanoparticles using carbon dioxide.
PMID 28449985
Abstract
Fish oil was encapsulated in hollow solid lipid micro- and nanoparticles formed from fully hydrogenated soybean oil (FHSO) using a novel green method based on atomization of supercritical carbon dioxide (SC-CO2)-expanded lipid. The highest fish oil loading efficiency (97.5%, w/w) was achieved at 50%, w/w, initial fish oil concentration. All particles were spherical and in the dry free-flowing form; however, less smooth surface with wrinkles was observed when the initial fish oil concentration was increased up to 50%. With increasing initial fish oil concentration, melting point of the fish oil-loaded particles shifted to lower onset melting temperatures, and major polymorphic form transformed from α to β and/or β'. Oxidative stability of the loaded fish oil was significantly increased compared to the free fish oil (p<0.05). This innovative method forms free-flowing powder products that are easy-to-use solid fish oil formulation, which makes the handling and storage feasible and convenient.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
247502
Cumene hydroperoxide, technical grade, 80%
C9H12O2
A88255
p-Anisidine, ≥99%
C7H9NO