EMAIL THIS PAGE TO A FRIEND
Journal of comparative pathology
Vaccination against Louping Ill Virus Protects Goats from Experimental Challenge with Spanish Goat Encephalitis Virus.
PMID 28457486
Abstract
Spanish goat encephalitis virus (SGEV) is a recently described member of the genus Flavivirus belonging to the tick-borne encephalitis group of viruses, and is closely related to louping ill virus (LIV). Naturally acquired disease in goats results in severe, acute encephalitis and 100% mortality. Eighteen goats were challenged subcutaneously with SGEV; nine were vaccinated previously against LIV and nine were not. None of the vaccinated goats showed any clinical signs of disease or histological lesions, but all of the non-vaccinated goats developed pyrexia and 5/9 developed neurological clinical signs, primarily tremors in the neck and ataxia. All non-vaccinated animals developed histological lesions restricted to the central nervous system and consistent with a lymphocytic meningomyeloencephalitis. Vaccinated goats had significantly (P <0.003) greater concentrations of serum IgG and lower levels of IgM (P <0.0001) compared with unvaccinated animals. SGEV RNA levels were below detectable limits in the vaccinated goats throughout the experiment, but increased rapidly and were significantly (P <0.0001) greater 2-10 days post challenge in the non-vaccinated group. In conclusion, vaccination of goats against LIV confers highly effective protection against SGEV; this is probably mediated by IgG and prevents an increase in viral RNA load in serum such that vaccinated animals would not be an effective reservoir of the virus.