EMAIL THIS PAGE TO A FRIEND
Brain research bulletin
The protective effect of astaxanthin on learning and memory deficits and oxidative stress in a mouse model of repeated cerebral ischemia/reperfusion.
PMID 28479214
Abstract
Oxidative stress has been implicated in the pathogenesis of neurodegenerative disorders, such as vascular cognitive impairment (VCI). The present study was performed to investigate the potential neuroprotective effect of the antioxidant astaxanthin (ATX) in a mouse model of VCI. VCI was induced in male ICR mice by repeated occlusion of the bilateral common carotid artery, leading to repeated cerebral ischemia/reperfusion (IR) injury. After surgery, the mice received ATX or an equal volume of vehicle by daily intragastric administration for 28days. The results showed that ATX treatment ameliorated learning and memory deficits after repeated cerebral IR. ATX administration rescued the number of surviving pyramidal neurons in the CA1 and CA3 regions. The concentration of malondialdehyde was decreased, and the levels of reduced glutathione and superoxide dismutase in the hippocampus were increased. Electron microphotography revealed that damage to the ultrastructure of neurons was also reduced by ATX administration. In addition, the expression levels of Cytochrome C (Cyt C), cleaved Caspase-3 and Bax were lower and the expression of Bcl-2 was higher compared to control IR mice. Our findings demonstrate that ATX is able to suppresse learning and memory impairment caused by repeated cerebral IR and that this effect is associated with attenuation of oxidative stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89640
Toluidine Blue, for microscopy (Hist., Vit.)
C15H16ClN3S · 0.5ZnCl2