EMAIL THIS PAGE TO A FRIEND
Cell reports
A Chimeric Egfr Protein Reporter Mouse Reveals Egfr Localization and Trafficking Inxa0Vivo.
PMID 28494873
Abstract
EGF receptor (EGFR) is a critical signaling node throughout life. However, it has not been possible to directly visualize endogenous Egfr in mice. Using CRISPR/Cas9 genome editing, we appended a fluorescent reporter to the C terminus of the Egfr. Homozygous reporter mice appear normal and EGFR signaling is intact inxa0vitro and inxa0vivo. We detect distinct patterns of Egfr expression in progenitor and differentiated compartments in embryonic and adult mice. Systemic delivery of EGF or amphiregulin results in markedly different patterns of Egfr internalization and trafficking in hepatocytes. In the normal intestine, Egfr localizes to the crypt rather than villus compartment, expression is higher in adjacent epithelium than in intestinal tumors, and following colonic injury expression appears in distinct cell populations in the stroma. This reporter, under control of its endogenous regulatory elements, enables inxa0vivo monitoring of the dynamics of Egfr localization and trafficking in normal and disease states.