EMAIL THIS PAGE TO A FRIEND
FEMS yeast research
CRISPR/Cas system for yeast genome engineering: advances and applications.
PMID 28505256
Abstract
The methods based on the Clustered Regularly Interspaced Short Palindromic Repeats (CRISPR)/CRISPR-associated (Cas) system have quickly gained popularity for genome editing and transcriptional regulation in many organisms, including yeast. This review aims to provide a comprehensive overview of CRISPR application for different yeast species: from basic principles and genetic design to applications.