EMAIL THIS PAGE TO A FRIEND
Journal of assisted reproduction and genetics
Erratum to: Introducing precise genetic modifications into human 3PN embryos by CRISPR/Cas-mediated genome editing.
PMID 28508226