EMAIL THIS PAGE TO A FRIEND
Journal of materials science. Materials in medicine
Fabricating poly(1,8-octanediol citrate) elastomer based fibrous mats via electrospinning for soft tissue engineering scaffold.
PMID 28510114
Abstract
Poly(1,8-octanediol citrate) (POC) is a recently developed biodegradable crosslinked elastomer that possesses good cytocompatibility and matchable mechanical properties to soft tissues. However, the thermosetting characteristic reveals a big challenge to manufacture its porous scaffold. Herein, POC elastomer was electrospun into fiber mat using poly(L-lactic acid) (PLLA) as a spinnable carrier. The obtained POC/PLLA fiber mats were characterized by scanning electron microscopy (SEM), dynamic mechanical analysis (DMA), uniaxial tensile test, static-water-contact-angle, thermal analysis, in vitro degradation and biocompatibility test. It was found that the fibrous structure could be formed so long as the POC pre-polymer's content was no more than 50 wt%. The presence of elastic POC component not only strengthened the fiber mats but also toughened the fiber mats. The hydrophilicity of 50/50 fiber mat significantly improved. In vitro degradation rate of POC based fiber mats was much faster than that of pure PLLA. Cyto- and histo-compatibility tests confirmed that the POC/PLLA fiber mats had good biocompatibility for potential applications in soft tissue engineering.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4562
5-(N,N-Dimethyl)amiloride hydrochloride
C8H12ClN7O · HCl