EMAIL THIS PAGE TO A FRIEND
Acta cirurgica brasileira
Propofol attenuates cytokine-mediated upregulation of expression of inducible nitric oxide synthase and apoptosis during regeneration post-partial hepatectomy.
PMID 28591369
Abstract
To determine the effects of propofol and ketamine anesthesia on liver regeneration in rats after partial hepatectomy (PHT). Male Wistar albino rats were assigned randomly to four groups of 10. Anesthesia was induced and maintained with propofol in groups 1 and 2, and with ketamine in groups 3 and 4. PHT was undertaken in groups 1 and 3. Rats in groups 2 and 4 (control groups) underwent an identical surgical procedure, but without PHT. At postoperative day-5, rats were killed. Regenerated liver was removed, weighed, and evaluated (by immunohistochemical means) for expression of inducible nitric oxide synthase (iNOS), endothelial NOS (eNOS), apoptosis protease-activating factor (APAF)-1, and proliferating cell nuclear antigen (PCNA). Also, blood samples were collected for measurement of levels of tumor necrosis factor (TNF)-α and interleukin (IL)-6. Between groups 2 and 4, there were no differences in tissue levels of iNOS, eNOS, and APAF-1 or plasma levels of TNF-α and IL-6. eNOS expression was similar in group 1 and group 3. Expression of iNOS and APAF-1 was mild-to-moderate in group 1, but significantly higher in group 3. Groups 1 and 3 showed an increase in PCNA expression, but expression in both groups was comparable. Plasma levels of TNF-α and IL-6 increased to a lesser degree in group 1 than in group 3. Propofol, as an anesthetic agent, may attenuate cytokine-mediated upregulation of iNOS expression and apoptosis in an animal model of liver regeneration after partial hepatectomy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
155209
Pyrrolidone hydrotribromide, 97%
C12H22Br3N3O3