EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Sodium butyrate induces new gene expression in Friend erythroleukemic cells.
PMID 285931
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19364
Sodium butyrate, ≥99.0% (GC)
C4H7NaO2