EMAIL THIS PAGE TO A FRIEND
Scientific reports
Specific ablation of mouse Fam20C in cells expressing type I collagen leads to skeletal defects and hypophosphatemia.
PMID 28620244
Abstract
FAM20C mutations in humans cause Raine syndrome and our previous studies showed that global inactivation of mouse Fam20C led to bone and dental defects. By crossbreeding 2.3  kb Col 1a1-Cre mice with Fam20C (flox/flox) mice, we created 2.3  kb Col 1a1-Cre;Fam20C (foxl/flox) (cKO) mice, in which Fam20C was inactivated in cells expressing Type I collagen. This study showed that the long bones of cKO mice were shorter and had a lower level of mineralization compared to the normal mice. The collagen fibrils in Fam20C-deficient bone were disorganized and thicker while the growth plate cartilage in cKO mice was disorganized and wider compared to the normal mice. The Fam20C-deficient bone had a lower level of dentin matrix protein 1, and higher levels of osteopontin and bone sialoprotein than the normal. The blood of cKO mice had an elevated level of fibroblast growth factor 23 and reduced level of phosphorus. These findings indicate that inactivation of Fam20C in cells expressing type I collagen led to skeletal defects and hypophosphatemia. The altered levels of dentin matrix protein 1 and osteopontin in Fam20C-deficient bone may be significant contributors to the mineralized tissue defects in human patients and animals suffering from the functional loss of FAM20C.