EMAIL THIS PAGE TO A FRIEND
Drug intelligence & clinical pharmacy
Astemizole: a long-acting, nonsedating antihistamine.
PMID 2892659
Abstract
Astemizole is a long-acting, highly selective histamine1-receptor antagonist with minimal central and anticholinergic effects. Comparison studies have shown astemizole to be equal or superior to currently available antihistamines, beclomethasone nasal spray, and cromolyn sodium in relieving allergic symptoms of seasonal and perennial allergic rhinitis. Other uses include treatment of allergic conjunctivitis and chronic urticaria. Astemizole is not as effective for treatment of acute allergic symptoms because of its delayed onset of action. Astemizole and its active metabolite, desmethylastemizole, have long elimination half-lives permitting once-daily dosing. The incidence of sedation is lower than with conventional antihistamines, but increased appetite and weight gain do occur. Astemizole should be useful for both maintenance and prophylactic therapy in patients with chronic allergic conditions who cannot tolerate the sedative or anticholinergic effects of conventional antihistamines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2861
Astemizole, ≥98% (HPLC)
C28H31FN4O