EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A newly synthesized selective casein kinase I inhibitor, N-(2-aminoethyl)-5-chloroisoquinoline-8-sulfonamide, and affinity purification of casein kinase I from bovine testis.
PMID 2925675
Abstract
When screening various isoquinolinesulfonamide compounds which we synthesized, CKI-7, N-(2-amino-ethyl)-5-chloroisoquinoline-8-sulfonamide, was found to have a potent inhibitory action against casein kinase I and a much weaker effect on casein kinase II and other protein kinases. Kinetic analysis indicated that CKI-7 inhibited casein kinase I competitively with respect to ATP and that the Ki values were 8.5 microM for casein kinase I and 70 microM for casein kinase II. An affinity chromatography absorbent was synthesized by coupling CKI-8 (1-(5-chloroisoquinoline-8-sulfonyl], a derivative of CKI-7, to cyanogen bromide-activated Sepharose 4B. Partially purified casein kinase I from bovine testis was subjected to affinity chromatography. Analysis of the purified casein kinase I by polyacrylamide gel electrophoresis in the presence of sodium dodecyl sulfate revealed a single band with molecular weight 37,000. These newly synthesized compounds, CKI-7 and CKI-8, should serve as useful tools for elucidating the biological significance of casein kinase I-mediated reactions.