EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Solid-phase enzyme-receptor assay (SPERA): a competitive-binding assay for glycopeptide antibiotics of the vancomycin class.
PMID 2931213
Abstract
A solid-phase enzyme-receptor assay (SPERA) has been developed for glycopeptide antibiotics of the vancomycin class such as teicoplanin, vancomycin, ristocetin, avoparcin, actaplanin, A-47934, A-41030, and A-35512-B. The assay exploits the mechanism of most action of these antibiotics, which is based on their interaction with acyl-D-alanyl-D-alanine, a constituent of the walls of most growing bacterial cells. The antibiotics and enzyme-labeled teicoplanin compete for a synthetic analog of the biological receptor, albumin-epsilon-aminocaproyl-D-alanyl-D-alanine. The various antibiotics produced different competition curves, 50% displacement being obtained with antibiotic concentrations ranging from 0.04 to 4 mg/L, vancomycin and actaplanin being the weakest and strongest competitors, respectively. For teicoplanin in human serum the intra-assay CV was 7.2%, the interassay CV was 11.2%, and the analytical recovery 94%. Teicoplanin concentrations obtained by SPERA (chi) correlated well with those obtained by microbiological assay (y): y = 1.03 chi + 0.053 (r = 0.943; n = 60). We conclude that SPERA is a powerful tool for identification and quantitative detection of glycopeptide antibiotics, even in complex media.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3435 D-Alanine–Agarose, saline suspension