EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Restriction and modification enzymes and their recognition sequences.
PMID 2987885
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R5257
Spe I from Sphaerotilus sp., Restriction Enzyme