EMAIL THIS PAGE TO A FRIEND
Biomaterials
Structural analysis of hydroxyapatite coatings on titanium.
PMID 3011139
Abstract
Hydroxyapatite from two sources was electrophoretically deposited onto flat titanium plate material. Depending upon the deposition conditions various changes in the structure of the ceramic were identified. A well-adhering Ti-P compound was present at the interface. Hydroxyapatite oxygenated to various degrees and tetracalcium phosphate were reproducibly formed in the coating.