EMAIL THIS PAGE TO A FRIEND
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes
Radiopharmaceuticals labelled with bromine isotopes.
PMID 3021671