EMAIL THIS PAGE TO A FRIEND
Contributions to nephrology
Calcium carbonate and magnesium hydroxide in the prevention of renal osteodystrophy or the demise of aluminum toxicity in uremia. Analysis of 5 years experience.
PMID 3053039