EMAIL THIS PAGE TO A FRIEND
Veterinary and human toxicology
Toxicology of thiram (tetramethylthiuram disulfide): a review.
PMID 3055654
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T24201
Tetramethylthiuram disulfide, 97%
C6H12N2S4
45689
Thiram, PESTANAL®, analytical standard
C6H12N2S4