EMAIL THIS PAGE TO A FRIEND
Progress in brain research
Striatal grafts in the ibotenic acid-lesioned neostriatum: functional studies.
PMID 3073422
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2765
Ibotenic acid, ~95%, solid
C5H6N2O4