EMAIL THIS PAGE TO A FRIEND
Nature
Isolation of complementary DNA clones encoding the human lymphocyte glycoprotein T1/Leu-1.
PMID 3093892
Abstract
The T1/Leu-1/CD5 molecule, a human T-cell surface glycoprotein of relative molecular mass (Mr) 67,000, has been implicated in the proliferative response of activated T cells and in T-cell helper function. A similar involvement in T-cell proliferation has been reported for Ly-1, the murine homologue of T1. Here we report the complete amino-acid sequence of the T1 precursor molecule deduced from complementary DNA clones. The protein contains a classical signal peptide; a 347-amino-acid extracellular segment; a transmembrane region; and a 93-amino-acid intracellular segment. The extracellular segment contains many cysteine residues and is composed of two related structural domains separated by a proline/threonine-rich region. The T1 molecule has structural features characteristic of other receptor molecules.