EMAIL THIS PAGE TO A FRIEND
Heart & lung : the journal of critical care
Flecainide acetate: critical care administration guidelines.
PMID 3123423
Abstract
Flecainide may play an important role in the treatment of ventricular arrhythmias because of its convenient dosing regimen and high rate of efficacy in controlling ventricular ectopic activity. In stable patients with normal or mildly reduced left ventricular function, flecainide has proved to be promising. In those patients with severe left ventricular dysfunction, lethal arrhythmias, and preexisting conduction system disease, caution must be exercised.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0120000
Flecainide acetate, European Pharmacopoeia (EP) Reference Standard
C17H20F6N2O3 · C2H4O2
F6777
Flecainide acetate salt
C17H20F6N2O3 · C2H4O2