EMAIL THIS PAGE TO A FRIEND
Journal of immunological methods
Depletion of mycoplasma from infected cell lines by limiting dilution in 6-methylpurine deoxyriboside.
PMID 3127473
Abstract
Cells contaminated with mycoplasma were cloned in the presence of 6-methylpurine deoxyriboside. The results showed that a limiting dilution in the culture medium containing the chemical efficiently depletes mycoplasma providing the cells are sensitive to 0.6-1.2 microM 6-methylpurine.