EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Use of antiadhesive peptide and swainsonine to inhibit metastasis.
PMID 3149881
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9263
Swainsonine, synthetic
C8H15NO3
S8195
Swainsonine, from Metarrhizium anisopliae, ≥98% (TLC)
C8H15NO3