EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
High-resolution two-dimensional electrophoresis of plant proteins.
PMID 3189777
Abstract
A technique for the analysis of plant proteins from seed, leaf, root, and coleoptile tissues by high resolution two-dimensional electrophoresis is described. This technique is based primarily on the procedure of P. O'Farrell (1975, J. Biol. Chem. 250, 4007-4021); however, a number of improvements and simplifications have been introduced. We have found that resolution of polypeptides from a range of plant tissues is improved if the concentrations of nonionic detergent and ampholytes used in the isoelectric focusing (IEF) step are increased to 4 and 5% (w/v), respectively. Further increase in the concentrations of these two components results in gels of decreased resolution and low mechanical strength. We have also found that substitution of n-octyl beta-D-glucopyranoside or 3-[(cholamidopropyl)dimethylammonio]-1-propanesulfonate for Triton X-100 or Nonidet-P40 in the IEF dimension significantly increases the resolution of polypeptides in these gels. This technique also allows minor polypeptide differences between closely related cultivars of plants to be identified.