EMAIL THIS PAGE TO A FRIEND
Clinical pediatrics
5-fluorocytosine: a brief review.
PMID 321173
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0175000
Flucytosine, European Pharmacopoeia (EP) Reference Standard
C4H4FN3O