EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Methylazoxymethanol treatment of fetal rats results in abnormally dense noradrenergic innervation of neocortex.
PMID 32620
Abstract
A single injection of methylazoxymethanol in pregnant rats at 15 days of gestation results in severe cortical atrophy in the offspring. In the adult offspring, the neurochemical markers for the cortical gamma-aminobutyric acid-containing neurons are severely reduced, whereas the noradrenergic markers are minimally altered. Immunohistofluorescence microscopy demonstrates a marked increase in the density of noradrenergic axons which have an abnormal pattern of distribution in the atrophic cortex. The results suggest that the central noradrenergic neurons determine the number of axons to be formed early in brain development, but local factors in the terminal field regulate the ultimate distribution of the noradrenergic axons.