EMAIL THIS PAGE TO A FRIEND
Revista espanola de las enfermedades del aparato digestivo
[Liver lesion caused by cyanamide. Study of a case and review of the bibliography].
PMID 3291022
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
187364
Cyanamide, 99%
CH2N2
C87908
Cyanamide solution, 50 wt. % in H2O
CH2N2