EMAIL THIS PAGE TO A FRIEND
Histopathology
A simple guide to the terminology and application of leucocyte monoclonal antibodies.
PMID 3294157
Abstract
This review aims to provide a simple guide and quick reference to the terminology and diagnostic applications of leucocyte monoclonal antibodies. The differentiation cluster terminology, where applicable, is used throughout.