EMAIL THIS PAGE TO A FRIEND
Pediatrics
Pimozide treatment of tic and Tourette disorders.
PMID 3295739
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1793 Pimozide
C28H29F2N3O