EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Perspectives on the mechanism of action of the splenic toxicity of aniline and structurally-related compounds.
PMID 3305247
Abstract
Aniline and several structurally-related aromatic amines produce spleen tumours in rats given high doses of compound in 2-year bioassay studies. Evaluation of the pathogenesis of the splenic lesions and characterization of the disposition of radiolabelled aniline in animals suggests that the spleen tumours may be a secondary response resulting from chemically-mediated erythrocyte toxicity. It is proposed that compound-derived toxicity to erythrocytes results in scavenging of damaged red blood cells by the spleen, initiating a series of events which may contribute to the development of spleen tumours. These events potentially include (i) specific accumulation of the parent compound or toxic metabolite(s) carried to the spleen by erythrocytes; (ii) deposition of erythrocytic debris, particularly iron, which may catalyse tissue-damaging free-radical reactions; and (iii) induction of splenic hyperplasia resulting from erythrocyte overload. Linkage of the splenic tumorigenicity of these aromatic amines to an initial toxic event in the erythrocyte suggests that the carcinogenicity of such compounds may be determined by a definable threshold dose, i.e. the events leading to the carcinogenicity are not initiated until the capacity of the red blood cell to cope with the toxic insult is exceeded.