EMAIL THIS PAGE TO A FRIEND
Radiology
Multiple cerebral metastases: detectability with Gd-DTPA-enhanced MR imaging.
PMID 3317495
Abstract
Sixteen patients with suspected cerebral metastases were studied with magnetic resonance (MR) imaging before and after the intravenous administration of 0.1 mmol/kg of gadolinium diethylenetriaminepenta-acetic acid. The images were interpreted blindly by two neuroradiologists; all clinical, radiologic (computed tomographic and MR imaging), and pathologic data were reviewed to arrive at a final "best diagnosis," which was then compared with the prior blinded interpretations. Of seven patients found to have multiple metastases, six (86%) had at least one tumor nodule depicted by postinfusion MR imaging that was missed by one or both observers on review of preinfusion images alone. Lesions missed on preinfusion studies were usually small nodules hidden by or not detected next to regions of high-signal edema thought to be related to the adjacent tumor nodule. The authors believe that contrast enhancement improves detection of metastatic foci with MR imaging and that the findings indicate broader implications for the detection of multiple lesions from other causes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
381667
Diethylenetriaminepentaacetic acid gadolinium(III) dihydrogen salt hydrate, 97%
C14H20GdN3O10 · xH2O