EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica et toxicologica
The pharmacology of bumetanide.
PMID 331869
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3023
Bumetanide, ≥98%
C17H20N2O5S
B1158000
Bumetanide, European Pharmacopoeia (EP) Reference Standard
C17H20N2O5S