EMAIL THIS PAGE TO A FRIEND
Rinsho shinkeigaku = Clinical neurology
[Report on an autopsy of a cancer patient with Carmofur (HCFU) leukoencephalopathy].
PMID 3329075
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1494
Carmofur, ≥98% (HPLC), powder
C11H16FN3O3