EMAIL THIS PAGE TO A FRIEND
Proteins
Cloning, expression, and nucleotide sequence of livR, the repressor for high-affinity branched-chain amino acid transport in Escherichia coli.
PMID 3329726
Abstract
The livR gene encoding the repressor for high-affinity branched-chain amino acid transport in Escherichia coli has been cloned from a library prepared from the episome F106. The inserted DNA fragment from the initial cloned plasmid, pANT1, complemented two independent, spontaneously derived, regulatory mutations. Subcloning as well as the creation of deletions with Bal31 exonuclease revealed that the entire regulatory region is contained within a 1.1-kb RsaI-SalI fragment. Expression of the pANT plasmids in E. coli minicells showed that the regulatory region encodes one detectable protein with an apparent molecular weight of 21,000. DNA sequencing revealed one open reading frame of 501 bp encoding a protein with a calculated MW of 19,155. The potential secondary structure of the regulatory protein has been predicted and it suggests that the carboxy terminus may fold into three consecutive alpha helices. These results suggest that the livR gene encodes a repressor which plays a role in the regulation of expression of the livJ and the livK transport genes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6514
3-(Trimethoxysilyl)propyl methacrylate, ≥98%
C10H20O5Si