EMAIL THIS PAGE TO A FRIEND
Cancer chemotherapy and pharmacology
DNA repair following incorporation of 5-fluorouracil into DNA of mouse bone marrow cells.
PMID 3359556
Abstract
5-Fluorouracil (FUra) was previously demonstrated to be incorporated into DNA at cytotoxic concentrations in mouse bone marrow cells. Subsequently, we showed that under these conditions FUra exhibited a time-dependent removal from DNA accompanied by a decrease in DNA strand length. In the present study we utilized hydroxyurea to inhibit semiconservative DNA synthesis while monitoring DNA repair by measuring the incorporation of [3H]dThd into double-stranded DNA (DNAds), which can be separated from DNA at the replication fork (DNAss) by benzoylated-naphthoylated-DEAE cellulose. Under these conditions we assessed DNA repair in cells that had previously been exposed for 1 h to varying cytotoxic concentrations of FUra. The results demonstrate an increase in labelling of DNAds with increasing FUra concentrations, with no appreciable increase in incorporation of label into DNAss. In conclusion, this study demonstrates that DNA repair occurs following incorporation of FUra. The failure to repair DNA damage at higher FUra concentrations may have a role in the cytotoxicity of this drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B6385
Benzoylated Naphthoylated DEAE–Cellulose, medium