EMAIL THIS PAGE TO A FRIEND
Cold Spring Harbor symposia on quantitative biology
Erythropoietin: gene cloning, protein structure, and biological properties.
PMID 3472747
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11120166001 Erythropoietin, human (hEPO), recombinant (CHO cells)