EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
B-cell-stimulatory factor 2 (beta 2 interferon) functions as a second signal for interleukin 2 production by mature murine T cells.
PMID 3499611
Abstract
Purified peripheral murine T cells, in the presence of concanavalin A, can be activated to produce interleukin 2 (IL-2) through stimulation either with a previously described murine lymphokine designated T cell-activating factor (TAF) or with a cloned human lymphokine that has been called beta 2 interferon, B-cell-stimulatory factor 2, hybridoma growth factor, inducible 26-kDa protein, or hematopoietic colony-stimulating factor 309 by different investigators. We and others propose the designation interleukin 6 (IL-6) for the latter molecule. Our experiments demonstrate that either murine TAF or human IL-6 can restore the ability of purified T cells to proliferate in response to Con A or antibodies against the T-cell antigen receptor. Most if not all of the proliferation can be blocked by antibodies against the alpha chain of the IL-2 receptor. Furthermore, highly purified CD8- T cells can be activated by IL-6 in the presence of Con A to secrete IL-2. We propose that IL-6 and murine TAF are important "second signals" in primary antigen-receptor-dependent T-cell activation. Whether or not murine TAF is a homologue of human IL-6 remains to be determined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11138600001 Interleukin-6, human (hIL-6), recombinant (E. coli)