EMAIL THIS PAGE TO A FRIEND
The Analyst
Automated flow injection spectrophotometric determination of zinc using zincon: applications to analysis of waters, alloys and insulin formulations.
PMID 3544955
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
96440
Zincon monosodium salt, for spectrophotometric det. of Cu, Zn
C20H15N4NaO6S