EMAIL THIS PAGE TO A FRIEND
Medicinal research reviews
The chemistry, pharmacology, and clinical applications of qinghaosu (artemisinin) and its derivatives.
PMID 3550324
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7439
Dihydroartemisinin, analytical standard, mixture of α and β isomers
C15H24O5