EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of the carcinogenic risk of chemicals to man
N-nitroso-n-methylurea.
PMID 355099
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1517
N-Nitroso-N-methylurea, ISOPAC®
C2H5N3O2
N4766
N-Nitroso-N-methylurea, Bulk package
C2H5N3O2