EMAIL THIS PAGE TO A FRIEND
Nature
Antimicrobial peptides. A family of wound healers.
PMID 3614352
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7402
Magainin II, ≥97% (HPLC)
C114H180N30O29S