EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Efficient stereospecific synthesis of no-carrier-added 2-[18F]-fluoro-2-deoxy-D-glucose using aminopolyether supported nucleophilic substitution.
PMID 3712040
Abstract
An aminopolyether mediated synthesis of fluorine-18 (18F) 2-fluoro-2-deoxy-D-glucose (FDG) has been developed. The nucleophilic fluorination with accelerator-produced [18F]fluoride works at the no-carrier-added level and gives epimerically pure 2-18FDG with an uncorrected radiochemical yield of a maximum 50% in a synthesis time of approximately 50 min from EOB.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1568
Mannose triflate, For PET imaging, ≥98% (TLC)
C15H19F3O12S