EMAIL THIS PAGE TO A FRIEND
Life sciences
Biochemical pharmacology of GABAergic agonists.
PMID 37392
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G002
Isoguvacine hydrochloride, solid
C6H9NO2 · HCl