EMAIL THIS PAGE TO A FRIEND
The Journal of nursing administration
Tackling writer's block. Suggestions for self-modification.
PMID 3845971
Abstract
Writing is an important and pervasive part of every administrator's duties, and problems with writer's block and procrastination can impede one's productivity. This article describes behavioral assessment and intervention methods (e.g., task analysis, reinforcement analysis, time-structuring, charting, and deposit contracting) that an individual can use to modify his or her writing habits.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
327204 Paraffin wax, mp 53-57 °C (ASTM D 87)