EMAIL THIS PAGE TO A FRIEND
Nature
Human transforming growth factor-beta complementary DNA sequence and expression in normal and transformed cells.
PMID 3861940
Abstract
The partial amino-acid sequence of purified human transforming growth factor-beta (TGF-beta) was used to identify a series of cDNA clones encoding the protein. The cDNA sequence indicates that the 112-amino acid monomeric form of the natural TGF-beta homodimer is derived proteolytically from a much longer precursor polypeptide which may be secreted. TGF-beta messenger RNA is synthesized in various normal and transformed cells.
Related Materials