EMAIL THIS PAGE TO A FRIEND
Seikagaku. The Journal of Japanese Biochemical Society
[Polyacrylamide gel electrophoresis in the presence of lithium dodecyl sulfate. Problems and advantages].
PMID 3891886