EMAIL THIS PAGE TO A FRIEND
Drug intelligence & clinical pharmacy
New drug therapy for kidney stones: a review of cellulose sodium phosphate, acetohydroxamic acid, and potassium citrate.
PMID 3896714
Abstract
Kidney stones have an overall incidence of two to three percent in western countries. In many patients, the disease process is difficult to control and recurrence rates are high: 20 to 50 percent over the subsequent ten years. The pathogenesis and standard methods of treatment for the five major types of stones (i.e., calcium oxalate, struvite, calcium phosphate, uric acid, and cystine) are reviewed. Three new drugs are reviewed in the context of their roles in the selective treatment of kidney stones. Cellulose sodium phosphate (Calcibind) is a nonabsorbable ion-exchange resin with a limited indication for the treatment of calcium stones associated with absorptive hypercalciuria Type I. Acetohydroxamic acid (Lithostat) is an urease-inhibitor that is indicated as adjunctive therapy in patients with chronic urea-splitting urinary tract infections and struvite stones. Potassium citrate (Urocit) is an investigational agent that has clinical efficacy in patients with calcium oxalate and calcium phosphate stones who are hypocitraturic. In addition, potassium citrate is an alkalinizing agent that can be used in patients with uric acid stones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1006506
Acetohydroxamic acid, United States Pharmacopeia (USP) Reference Standard
C2H5NO2