EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
[Orotic acid].
PMID 3898151
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O2750
Orotic acid, ≥98% (titration), anhydrous
C5H4N2O4