EMAIL THIS PAGE TO A FRIEND
Journal of toxicology. Clinical toxicology
Sulindac hepatotoxicity: a case report and review.
PMID 3903180
Abstract
A 44 year old female, previously on propranolol, phenytoin and phenobarbital, developed hepatotoxicity while on sulindac and acetaminophen containing analgesic. A limited review of hepatotoxicity and drug interactions of sulindac is presented. The possible mechanism of hepatotoxicity and its treatment is suggested.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2180000
Sulindac, European Pharmacopoeia (EP) Reference Standard
C20H17FO3S